2-Hydroxy-4,5-dimethoxybenzaldehyde 98%
Supplier: Apollo Scientific
Synonyms: 4,5-Dimethoxy-2-hydroxybenzaldehyde
OR930296-1G OR930296-5G
APOSOR930296-1GEA 109 EUR
APOSOR930296-1G APOSOR930296-5G
2-Hydroxy-4,5-dimethoxybenzaldehyde 98%
2-Hydroxy-4,5-dimethoxybenzaldehyde
Formula: C₉H₁₀O₄
MW: 182.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00598182
CAS Number: 14382-91-3