2-Acetyl-1,3-cyclohexanedione 97%
Supplier: Apollo Scientific
Synonyms: 2-Acetylcyclohexane-1,3-dione
OR930355-10G OR930355-1G OR930355-5G
APOSOR930355-10GEA 430 EUR
APOSOR930355-10G APOSOR930355-1G APOSOR930355-5G
2-Acetyl-1,3-cyclohexanedione 97%
2-Acetyl-1,3-cyclohexanedione
Formula: C₈H₁₀O₃
MW: 154.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001586
CAS Number: 4056-73-9