1-(Dimethylamino)-2-methylpropan-2-ol 95%
Supplier: Apollo Scientific
OR930662-1G OR930662-5G
APOSOR930662-1GEA 77 EUR
APOSOR930662-1G APOSOR930662-5G
1-(Dimethylamino)-2-methylpropan-2-ol 95%
1-(Dimethylamino)-2-methylpropan-2-ol
Formula: C₆H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD11934288
CAS Number: 14123-48-9
UN: 1993
ADR: 3,III