7-Bromo-DL-tryptophan 97%
Supplier: Apollo Scientific
Synonyms: H-DL-Trp(7-Br)-OH, 2-Amino-3-(7-bromo-1H-indol-3-yl)propanoic acid
OR930993-1G OR930993-250MG
APOSOR930993-1GEA 710 EUR
APOSOR930993-1G APOSOR930993-250MG
7-Bromo-DL-tryptophan 97%
7-Bromo-DL-tryptophan
Formula: C₁₁H₁₁BrN₂O₂
MW: 283.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972047
CAS Number: 852391-45-8