2-(2,3-Dihydro-benzo[1,4]dioxin-2-yl)-ethylamine 95%
Supplier: Apollo Scientific
OR931424-1G OR931424-250MG
APOSOR931424-1GEA 864 EUR
APOSOR931424-1G APOSOR931424-250MG
2-(2,3-Dihydro-benzo[1,4]dioxin-2-yl)-ethylamine 95%
2-(2,3-Dihydro-benzo[1,4]dioxin-2-yl)-ethylamine
Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01659461
CAS Number: 87086-36-0