2,5-Dichloro-4-methoxypyridine 97%
Supplier: Apollo Scientific
OR931505-1G OR931505-250MG
APOSOR931505-1GEA 42.2 EUR
APOSOR931505-1G APOSOR931505-250MG
2,5-Dichloro-4-methoxypyridine 97%
2,5-Dichloro-4-methoxypyridine
Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185541
CAS Number: 1122090-95-2