2-Ethoxy-1-pyrroline 95%
Supplier: Apollo Scientific
OR931545-1G OR931545-25G OR931545-5G
APOSOR931545-1GEA 145 EUR
APOSOR931545-1G APOSOR931545-25G APOSOR931545-5G
2-Ethoxy-1-pyrroline 95%
2-Ethoxy-1-pyrroline
Formula: C₆H₁₁NO
MW: 113.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11618011
CAS Number: 931-46-4