2-Chlorocarbonyl-2-methyl-propionic acid ethyl ester 95%
Supplier: Apollo Scientific
Synonyms: Ethyl 3-chloro-2,2-dimethyl-3-oxopropanoate
OR931629-1G OR931629-250MG OR931629-5G
APOSOR931629-1GEA 197 EUR
APOSOR931629-1G APOSOR931629-250MG APOSOR931629-5G
2-Chlorocarbonyl-2-methyl-propionic acid ethyl ester 95%
2-Chlorocarbonyl-2-methyl-propionic acid ethyl ester
Formula: C₇H₁₁ClO₃
MW: 178.613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01961298
CAS Number: 64244-87-7