α,α,2,4-tetrachlorotoluene 98%
Supplier: Apollo Scientific
Synonyms: 2,4-Dichloro-1-(dichloromethyl)benzene
OR931922-5G
APOSOR931922-5GEA 68.3 EUR
APOSOR931922-5G
α,α,2,4-tetrachlorotoluene 98%
α,α,2,4-tetrachlorotoluene
Formula: C₇H₄Cl₄
MW: 229.919 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00043967
CAS Number: 134-25-8
UN: 3077
ADR: 9+EHS,III