4-(Dimethoxymethyl)-2-methoxypyrimidine 95%
Supplier: Apollo Scientific
OR931975-1G OR931975-250MG
APOSOR931975-1GEA 207 EUR
APOSOR931975-1G APOSOR931975-250MG
4-(Dimethoxymethyl)-2-methoxypyrimidine 95%
4-(Dimethoxymethyl)-2-methoxypyrimidine
Formula: C₈H₁₂N₂O₃
MW: 184.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275991
CAS Number: 193746-84-8