(7-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%
Supplier: Apollo Scientific
OR932051-1G OR932051-250MG
APOSOR932051-1GEA 322 EUR
APOSOR932051-1G APOSOR932051-250MG
(7-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%
(7-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol
Formula: C₉H₉ClN₂O
MW: 196.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18878718
CAS Number: 1340111-89-8