1,4-Dioxaspiro[4.5]decane-8-acetonitrile 95%
Supplier: Apollo Scientific
OR932219-1G OR932219-250MG
APOSOR932219-1GEA 632 EUR
APOSOR932219-1G APOSOR932219-250MG
1,4-Dioxaspiro[4.5]decane-8-acetonitrile 95%
1,4-Dioxaspiro[4.5]decane-8-acetonitrile
Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037894
CAS Number: 124499-37-2