2-tert-Butyl-5-chloro-1,1-dioxoisothiazol-3(2H)-one
Supplier: Apollo Scientific
Warning
Synonyms: 2-tert-Butyl-5-chloro-1,1-dioxo-1,2-thiazol-3-one
OR932241-250MG OR932241-1G
APOSOR932241-250MGEA 582 EUR
APOSOR932241-250MG APOSOR932241-1G
2-tert-Butyl-5-chloro-1,1-dioxoisothiazol-3(2H)-one
2-tert-Butyl-5-chloro-1,1-dioxoisothiazol-3(2H)-one
Formula: C₇H₁₀ClNO₃S
MW: 223.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029621
CAS Number: 850314-47-5