4-Aminopyrrolidin-2-one, 4-BOC protected 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (5-oxopyrrolidin-3-yl)carbamate
OR932307-1G OR932307-5G
APOSOR932307-1GEA 52.2 EUR
APOSOR932307-1G APOSOR932307-5G
4-Aminopyrrolidin-2-one, 4-BOC protected 97%
4-Aminopyrrolidin-2-one, 4-BOC protected
Formula: C₉H₁₆N₂O₃
MW: 200.235 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17677321
CAS Number: 1245648-84-3