Ethyl 4-methyl-1H-pyrazole-5-carboxylate 98%
Supplier: Apollo Scientific
OR932394-1G OR932394-5G
APOSOR932394-1GEA 166 EUR
APOSOR932394-1G APOSOR932394-5G
Ethyl 4-methyl-1H-pyrazole-5-carboxylate 98%
Ethyl 4-methyl-1H-pyrazole-5-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112107
CAS Number: 6076-12-6