(2-Chloro-4-methylphenyl)methanamine 96%
Supplier: Apollo Scientific
OR932412-1G
APOSOR932412-1GEA
140
EUR
APOSOR932412-1G
(2-Chloro-4-methylphenyl)methanamine 96%
(2-Chloro-4-methylphenyl)methanamine
Formula:
C₈H₁₀ClN MW: 155.625 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213616 CAS Number: 69957-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...