6-Chloro-1-methyl-1H-indazole 98%
Supplier: Apollo Scientific
OR932513-1G OR932513-250MG
APOSOR932513-1GEA 133 EUR
APOSOR932513-1G APOSOR932513-250MG
6-Chloro-1-methyl-1H-indazole 98%
6-Chloro-1-methyl-1H-indazole
Formula: C₈H₇ClN₂
MW: 166.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977521
CAS Number: 1210781-03-5