Ethyl 2-methylpyrimidine-5-carboxylate 95%
Supplier: Apollo Scientific
OR932690-1G OR932690-5G
APOSOR932690-1GEA 63.8 EUR
APOSOR932690-1G APOSOR932690-5G
Ethyl 2-methylpyrimidine-5-carboxylate 95%
Ethyl 2-methylpyrimidine-5-carboxylate
Formula: C₈H₁₀N₂O₂
MW: 166.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09863191
CAS Number: 2134-38-5