5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-1H-1,2,4-triazol-3(2H)-one
OR932692-1G
OR932692-250MG
OR932692-5G
APOSOR932692-1GEA
250
EUR
APOSOR932692-1G
APOSOR932692-250MG
APOSOR932692-5G
5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one
5-Amino-2,4-dihydro-3H-1,2,4-triazol-3-one
Formula:
C₂H₄N₄O MW: 100.079 g/mol Melting Pt: 286…290 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD23140949 CAS Number: 1003-35-6 |