Boc-Cys(stbu)-OH
Supplier: Apollo Scientific
Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(tert-butyldisulphanyl)propanoic acid
OR932786-1G OR932786-5G
APOSOR932786-1GEA 94.4 EUR
APOSOR932786-1G APOSOR932786-5G
Boc-Cys(stbu)-OH
Boc-Cys(stbu)-OH
Formula: C₁₂H₂₃NO₄S₂
Melting Pt: 110…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00084687
CAS Number: 30044-61-2