1-(3-Hydroxypyridin-2-yl)ethanone 95%
Supplier: Apollo Scientific
OR932930-1G
OR932930-5G
APOSOR932930-1GEA
149
EUR
APOSOR932930-1G
APOSOR932930-5G
1-(3-Hydroxypyridin-2-yl)ethanone 95%
1-(3-Hydroxypyridin-2-yl)ethanone
Formula:
C₇H₇NO₂ MW: 137.136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08062669 CAS Number: 13210-29-2 |