2-Chloro-1,5-naphthyridine 97%
Supplier: Apollo Scientific
OR933028-1G OR933028-250MG OR933028-5G
APOSOR933028-1GEA 178 EUR
APOSOR933028-1G APOSOR933028-250MG APOSOR933028-5G
2-Chloro-1,5-naphthyridine 97%
2-Chloro-1,5-naphthyridine
Formula: C₈H₅ClN₂
MW: 164.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189452
CAS Number: 7689-62-5