3-Bromo-1,5-dimethyl-1H-pyrazole 95%
Supplier: Apollo Scientific
OR933163-1G OR933163-250MG
APOSOR933163-1GEA 149 EUR
APOSOR933163-1G APOSOR933163-250MG
3-Bromo-1,5-dimethyl-1H-pyrazole 95%
3-Bromo-1,5-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991542
CAS Number: 5744-80-9