2-Amino-4'-hydroxyacetophenone hydrochloride
Supplier: Apollo Scientific
OR933236-1G OR933236-25G OR933236-5G
APOSOR933236-1GEA 58 EUR
APOSOR933236-1G APOSOR933236-25G APOSOR933236-5G
2-Amino-4'-hydroxyacetophenone hydrochloride
2-Amino-4'-hydroxyacetophenone hydrochloride
Formula: C₈H₉NO₂·ClH
MW: 187.624 g/mol
Melting Pt: 245…252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08436951
CAS Number: 19745-72-3