2,5-Dimethylhex-3-ene-2,5-diol 95%
Supplier: Apollo Scientific
OR933400-25G OR933400-100G
APOSOR933400-25GEA 87 EUR
APOSOR933400-25G APOSOR933400-100G
2,5-Dimethylhex-3-ene-2,5-diol 95%
2,5-Dimethylhex-3-ene-2,5-diol
Formula: C₈H₁₆O₂
MW: 144.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00968685
CAS Number: 927-81-1