2-Oxaspiro[3.5]nonane-6,8-dione
Supplier: Apollo Scientific
OR934727-100MG OR934727-1G OR934727-250MG
APOSOR934727-100MGEA 319 EUR
APOSOR934727-100MG APOSOR934727-1G APOSOR934727-250MG
2-Oxaspiro[3.5]nonane-6,8-dione
2-Oxaspiro[3.5]nonane-6,8-dione
Formula: C₈H₁₀O₃
MW: 154.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27988086
CAS Number: 1630907-24-2