2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride 95%
Supplier: Apollo Scientific
Synonyms: 2-Phenyl-1-(piperazin-1-yl)ethanone hydrochloride
OR934729-1G OR934729-250MG
APOSOR934729-1GEA 132 EUR
APOSOR934729-1G APOSOR934729-250MG
2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride 95%
2-Phenyl-1-(piperazin-1-yl)ethan-1-one hydrochloride
Formula: C₁₂H₁₆N₂O·ClH
MW: 240.729 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28040565
CAS Number: 502653-18-1