2-Propoxy-pyridine-3-carbaldehyde 97%
Supplier: Apollo Scientific
OR937480-1G OR937480-250MG
APOSOR937480-1GEA 319 EUR
APOSOR937480-1G APOSOR937480-250MG
2-Propoxy-pyridine-3-carbaldehyde 97%
2-Propoxy-pyridine-3-carbaldehyde
Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06804561
CAS Number: 865278-12-2