4,6-Dimethylnicotinonitrile 95%
Supplier: Apollo Scientific
OR938736-5G
APOSOR938736-5GEA 149 EUR
APOSOR938736-5G
4,6-Dimethylnicotinonitrile 95%
4,6-Dimethylnicotinonitrile
Formula: C₈H₈N₂
MW: 132.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234419
CAS Number: 6623-21-8