3-Methylbenzenesulphonamide 95%
Supplier: Apollo Scientific
OR938766-1G
OR938766-250MG
APOSOR938766-1GEA
224
EUR
APOSOR938766-1G
APOSOR938766-250MG
3-Methylbenzenesulphonamide 95%
3-Methylbenzenesulphonamide
Formula:
C₇H₉NO₂S MW: 171.217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04066695 CAS Number: 1899-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...