2-Methyl-2-phenylpropan-1-ol 95%
Supplier: Apollo Scientific
Synonyms: 2-Methyl-2-phenyl-1-propanol
OR938847-1G OR938847-250MG
APOSOR938847-1GEA 72.5 EUR
APOSOR938847-1G APOSOR938847-250MG
2-Methyl-2-phenylpropan-1-ol 95%
2-Methyl-2-phenylpropan-1-ol
Formula: C₁₀H₁₄O
MW: 150.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00506139
CAS Number: 2173-69-5