2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 95%
Supplier: Apollo Scientific
OR938848-1G
APOSOR938848-1GEA 52.2 EUR
APOSOR938848-1G
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 95%
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine
Formula: C₇H₈N₂S
MW: 152.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038619
CAS Number: 36267-71-7