4-Butylbenzyl alcohol 98%

Supplier: Apollo Scientific

Synonyms: (4-Butylphenyl)methanol

OR938874-1G OR938874-5G
APOSOR938874-1GEA 89.9 EUR
APOSOR938874-1G APOSOR938874-5G
4-Butylbenzyl alcohol 98%
4-Butylbenzyl alcohol
Formula: C₁₁H₁₆O
MW: 164.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010258
CAS Number: 60834-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR