2-Hydroxy-6-methylbenzaldehyde 95%
Supplier: Apollo Scientific
Synonyms: 6-Methylsalicylaldehyde
OR938926-1G OR938926-250MG
APOSOR938926-1GEA 162 EUR
APOSOR938926-1G APOSOR938926-250MG
2-Hydroxy-6-methylbenzaldehyde 95%
2-Hydroxy-6-methylbenzaldehyde
Formula: C₈H₈O₂
MW: 136.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16038308
CAS Number: 18362-36-2