2-Methyl-1-cyclohexanecarboxylic acid 95%
Supplier: Apollo Scientific
OR939113-1G OR939113-5G
APOSOR939113-1GEA 261 EUR
APOSOR939113-1G APOSOR939113-5G
2-Methyl-1-cyclohexanecarboxylic acid 95%
2-Methyl-1-cyclohexanecarboxylic acid
Formula: C₈H₁₄O₂
MW: 142.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074907
CAS Number: 56586-13-1