3,5-Dichloro-L-phenylalanine 97%
Supplier: Apollo Scientific
OR939204-1G OR939204-250MG OR939204-500MG
APOSOR939204-1GEA 236 EUR
APOSOR939204-1G APOSOR939204-250MG APOSOR939204-500MG
3,5-Dichloro-L-phenylalanine 97%
3,5-Dichloro-L-phenylalanine
Formula: C₉H₉Cl₂NO₂
MW: 234.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07372031
CAS Number: 13990-04-0