2-Methyl-2-cyclohexen-1-one 95%
Supplier: Apollo Scientific
Warning
OR939293-1G OR939293-250MG OR939293-500MG
APOSOR939293-1GEA 335 EUR
APOSOR939293-1G APOSOR939293-250MG APOSOR939293-500MG
2-Methyl-2-cyclohexen-1-one 95%
2-Methyl-2-cyclohexen-1-one
Formula: C₇H₁₀O
MW: 110.154 g/mol
Boiling Pt: 176…179 °C
Density: 0.9724 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00061069
CAS Number: 1121-18-2
UN: 1993
ADR: 3,III