2-Chloro-6-methylpyridin-4-amine 95%
Supplier: Apollo Scientific
OR939394-1G OR939394-250MG
APOSOR939394-1GEA 125 EUR
APOSOR939394-1G APOSOR939394-250MG
2-Chloro-6-methylpyridin-4-amine 95%
2-Chloro-6-methylpyridin-4-amine
Formula: C₆H₇ClN₂
MW: 142.586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235132
CAS Number: 79055-63-3