2,6-Dichloro-4-methoxyaniline 95%
Supplier: Apollo Scientific
OR939452-1G
APOSOR939452-1GEA 95.7 EUR
APOSOR939452-1G
2,6-Dichloro-4-methoxyaniline 95%
2,6-Dichloro-4-methoxyaniline
Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115065
CAS Number: 6480-66-6