2-Chloro-5-hydroxyisonicotinaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR939481-100MG
OR939481-250MG
APOSOR939481-100MGEA
98.6
EUR
APOSOR939481-100MG
APOSOR939481-250MG
2-Chloro-5-hydroxyisonicotinaldehyde 97%
2-Chloro-5-hydroxyisonicotinaldehyde
Formula:
C₆H₄ClNO₂ MW: 157.554 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189121 CAS Number: 1060804-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...