trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
Supplier: Apollo Scientific
Synonyms: trans-3-((tert-Butyldimethylsilyl)oxy)cyclobutanol
OR940901-1G OR940901-250MG
APOSOR940901-1GEA 578 EUR
APOSOR940901-1G APOSOR940901-250MG
trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
trans-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanol
Formula: C₁₀H₂₂O₂Si
MW: 202.366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23105931
CAS Number: 1408075-44-4