5-Chloro-6-methoxy-1-indanone 95%
Supplier: Apollo Scientific
Synonyms: 5-Chloro-6-methoxy-2,3-dihydro-1H-inden-1-one
OR940939-1G OR940939-250MG
APOSOR940939-1GEA 78.3 EUR
APOSOR940939-1G APOSOR940939-250MG
5-Chloro-6-methoxy-1-indanone 95%
5-Chloro-6-methoxy-1-indanone
Formula: C₁₀H₉ClO₂
MW: 196.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784346
CAS Number: 344305-70-0