2H-Benzo[e][1,3]oxazine-2,4(3H)-dione 95%
Supplier: Apollo Scientific
Synonyms: Carsalam
OR941085-100G OR941085-25G
APOSOR941085-100GEA 177 EUR
APOSOR941085-100G APOSOR941085-25G
2H-Benzo[e][1,3]oxazine-2,4(3H)-dione 95%
2H-Benzo[e][1,3]oxazine-2,4(3H)-dione
Formula: C₈H₅NO₃
MW: 163.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00600566
CAS Number: 2037-95-8