(S)-1-(Pyridin-3-yl)ethanamine 97%
Supplier: Apollo Scientific
Synonyms: (S)-1-(3-Pyridyl)ethylamine
OR941248-1G OR941248-250MG
APOSOR941248-1GEA 277 EUR
APOSOR941248-1G APOSOR941248-250MG
(S)-1-(Pyridin-3-yl)ethanamine 97%
(S)-1-(Pyridin-3-yl)ethanamine
Formula: C₇H₁₀N₂
MW: 122.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09256803
CAS Number: 27854-93-9