2-Chloro-4,5-dimethylaniline 95%
Supplier: Apollo Scientific
OR941440-1G OR941440-5G
APOSOR941440-1GEA 226 EUR
APOSOR941440-1G APOSOR941440-5G
2-Chloro-4,5-dimethylaniline 95%
2-Chloro-4,5-dimethylaniline
Formula: C₈H₁₀ClN
MW: 155.625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527213
CAS Number: 1585-13-3