6-Chloro-2-ethylbenzoxazole 95%
Supplier: Apollo Scientific
OR941742-1G
OR941742-250MG
APOSOR941742-1GEA
306
EUR
APOSOR941742-1G
APOSOR941742-250MG
6-Chloro-2-ethylbenzoxazole 95%
6-Chloro-2-ethylbenzoxazole
Formula:
C₉H₈ClNO MW: 181.619 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21641331 CAS Number: 1027019-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...