2-Chloro-3-methoxy-6-methylpyridine 95%
Supplier: Apollo Scientific
OR941863-1G OR941863-250MG
APOSOR941863-1GEA 301 EUR
APOSOR941863-1G APOSOR941863-250MG
2-Chloro-3-methoxy-6-methylpyridine 95%
2-Chloro-3-methoxy-6-methylpyridine
Formula: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD09260845
CAS Number: 204378-42-7