7-Methoxyimidazo[1,2-a]pyridine 95%
Supplier: Apollo Scientific
OR942142-1G OR942142-250MG
APOSOR942142-1GEA 104 EUR
APOSOR942142-1G APOSOR942142-250MG
7-Methoxyimidazo[1,2-a]pyridine 95%
7-Methoxyimidazo[1,2-a]pyridine
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185227
CAS Number: 342613-71-2