(2,5-Dimethoxy-1,4-phenylene)dimethanol 95%
Supplier: Apollo Scientific
OR942167-1G OR942167-5G
APOSOR942167-1GEA 154 EUR
APOSOR942167-1G APOSOR942167-5G
(2,5-Dimethoxy-1,4-phenylene)dimethanol 95%
(2,5-Dimethoxy-1,4-phenylene)dimethanol
Formula: C₁₀H₁₄O₄
MW: 198.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202848
CAS Number: 51829-43-7