Methyl 3-bromocyclobutane-1-carboxylate
Supplier: Apollo Scientific
OR942222-1G OR942222-5G
APOSOR942222-1GEA 68.3 EUR
APOSOR942222-1G APOSOR942222-5G
Methyl 3-bromocyclobutane-1-carboxylate
Methyl 3-bromocyclobutane-1-carboxylate
Formula: C₆H₉BrO₂
MW: 193.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20625735
CAS Number: 4935-00-6